Search
Search

Ccdc81| Gene Symbol: Ccdc81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162979.1 | NP_001156451.1 | 7 - 8 | 1255 | 73 | |
| GenBank mRNA | AK014876.1 | - | 7 - 8 | 1238 | 73 | |
| AK029544.1 | - | 7 - 8 | 1254 | 73 | ||
| BC127049.1 | - | 7 - 8 | 1242 | 73 | ||
| BC152307.1 | - | 7 - 8 | 1244 | 40062409 | 73 | |
| BC152308.1 | - | 7 - 8 | 1244 | 40062416 | 73 | |
Set Membership: |
|