Search
Search

Arhgap36| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081123.1 | NP_001074592.1 | 2 - 3 | 374 | 67 | |
| GenBank mRNA | AB073967.1 | - | 2 - 3 | 241 | 67 | |
| AK048167.1 | - | 2 - 3 | 262 | 67 | ||
| BC090672.1 | - | 2 - 3 | 345 | 30941825 | 67 | |
| BC145645.1 | - | 2 - 3 | 340 | 40130948 | 67 | |
Set Membership: |
|