Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01311611_m1
          See other STAG2 GE Assays ›
          Gene Symbol:
          Stag2
          Gene Name:
          stromal antigen 2
          Gene Aliases:
          9230105L23Rik, B230112I07Rik, SA-2, SAP2
          Chromosome Location:
          Chr.X: 42149322 - 42277186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01311611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stag2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077712.2 NP_001071180.1 2 - 3 407 114
          NM_001290713.1 NP_001277642.1 1 - 2 133 114
          NM_021465.4 NP_067440.3 2 - 3 267 114
          GenBank mRNA AK020316.1 - 2 - 3 212 114
          AK077318.1 - 2 - 3 279 114
          AK142460.1 - 2 - 3 377 114
          AK146718.1 - 2 - 3 267 114
          AK168899.1 - 2 - 3 267 114
          BC066041.1 - 2 - 3 149 114
          BU920543.1 - 2 - 3 407 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000398.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stag2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          cell cycle
          chromosome segregation
          sister chromatid cohesion
          mitotic nuclear division
          stem cell population maintenance
          negative regulation of DNA endoreduplication
          positive regulation of transcription from RNA polymerase II promoter
          cell division
          meiotic cell cycle
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00002657.mm.1 NM_001077712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002657.mm.1 TC0X00000398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002656.mm.1 NM_001077712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002656.mm.1 TC0X00000398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002664.mm.1 NM_001077712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002664.mm.1 NM_021465.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002664.mm.1 TC0X00000398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002665.mm.1 NM_001077712.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002665.mm.1 NM_021465.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002665.mm.1 TC0X00000398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002661.mm.1 NM_021465.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002661.mm.1 TC0X00000398.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline