Search
Search

Eftud1| Gene Symbol: Eftud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159672.1 | NP_001153144.1 | 12 - 13 | 1343 | 62 | |
| NM_175317.3 | NP_780526.2 | 12 - 13 | 1372 | 62 | ||
| GenBank mRNA | AK028676.1 | - | 12 - 13 | 1345 | 62 | |
| AK031647.1 | - | 12 - 13 | 1329 | 62 | ||
| AK049698.1 | - | 12 - 13 | 1331 | 62 | ||
| BC031852.1 | - | 6 - 7 | 618 | 62 | ||
| BC045616.1 | - | 12 - 13 | 1326 | 4983234 | 62 | |
Set Membership: |
|