Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01313038_m1
          See other PRSS55 GE Assays ›
          Gene Symbol:
          Prss55
          Gene Name:
          protease, serine 55
          Gene Aliases:
          4933401F05Rik
          Chromosome Location:
          Chr.14: 64075435 - 64085433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01313038_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss55
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081063.1 NP_001074532.1 4 - 5 663 95
          GenBank mRNA BC115562.2 - 4 - 5 664 40101914 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002228.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119613 NM_001081063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119613 AK016601.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137693 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86751 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119613 NM_001081063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119613 AK016601.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02868158 - -
          Whole Mouse Genome G4122A A_51_P235977 AK016601.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29665531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11485371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11688245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32974878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11349474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11688239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11364394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11681504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11250882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11688238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29665524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420806 - -
          Affymetrix HT MG-430 PM Array Plate 1453316_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420804 - -
          Mouse Genome 430 2.0 Array 1453316_PM_at - -
          Mouse Genome 430 2.0 Array 1453316_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71037_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71037_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400017252.mm.1 NM_001081063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017252.mm.1 NM_001081063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017252.mm.1 TC1400002228.mm.1 MTA TC
          Mouse Expression Set 430 1453316_at - -
          Mouse Expression Set 430 1453316_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453316_at - -
          Affymetrix HT Mouse Genome 430B Array 1453316_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 627557 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683640 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683640 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline