Search
Search

Parp4| Gene Symbol: Parp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145978.2 | NP_001139450.2 | 27 - 28 | 3453 | 60 | |
| GenBank mRNA | AK054229.1 | - | 27 - 28 | 3417 | 60 | |
| AK164513.1 | - | 27 - 28 | 3465 | 60 | ||
| AK172907.1 | - | 27 - 28 | 3385 | 60 | ||
Set Membership: |
|