Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01313813_m1
          See other EPHX2 GE Assays ›
          Gene Symbol:
          Ephx2
          Gene Name:
          epoxide hydrolase 2, cytoplasmic
          Gene Aliases:
          CEH, Eph2, SEH, sEP
          Chromosome Location:
          Chr.14: 66084372 - 66124522 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01313813_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ephx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271402.1 NP_001258331.1 3 - 4 519 62
          NM_001271403.1 NP_001258332.1 2 - 3 364 62
          NM_001271421.1 NP_001258350.1 2 - 3 414 62
          NM_007940.4 NP_031966.2 3 - 4 439 62
          GenBank mRNA AK002415.1 - 3 - 4 489 62
          AK142714.1 - 3 - 4 412 62
          AY098585.1 - 2 - 3 378 62
          BC015087.1 - 3 - 4 415 4218917 62
          EF597241.1 - 2 - 3 414 62
          L05781.1 - 3 - 4 352 62
          Z37107.1 - 3 - 4 369 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002266.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ephx2

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin production involved in inflammatory response
          lipid metabolic process
          inflammatory response
          metabolic process
          response to toxic substance
          positive regulation of gene expression
          dephosphorylation
          sensory perception of pain
          aromatic compound catabolic process
          cholesterol homeostasis
          linoleic acid metabolic process
          positive regulation of blood pressure
          stilbene catabolic process
          phospholipid dephosphorylation
          regulation of cholesterol metabolic process
          magnesium ion binding
          catalytic activity
          epoxide hydrolase activity
          receptor binding
          toxic substance binding
          hydrolase activity
          phosphatase activity
          10-hydroxy-9-(phosphonooxy)octadecanoate phosphatase activity
          lipid phosphatase activity
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400017504.mm.1 NM_001271421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017504.mm.1 TC1400002266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017502.mm.1 NM_001271402.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017502.mm.1 NM_001271403.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017502.mm.1 NM_001271421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017502.mm.1 NM_007940.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017502.mm.1 TC1400002266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017505.mm.1 NM_001271403.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017505.mm.1 TC1400002266.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0