Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01313910_m1
          See other SDR39U1 GE Assays ›
          Gene Symbol:
          Sdr39u1
          Gene Name:
          short chain dehydrogenase/reductase family 39U, member 1
          Gene Aliases:
          2310014G06Rik
          Chromosome Location:
          Chr.14: 55897285 - 55900281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01313910_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdr39u1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082975.1 NP_001076444.1 4 - 5 351 81
          GenBank mRNA AK009339.1 - 4 - 5 331 81
          AK049709.1 - 4 - 5 345 81
          BC087941.1 - 4 - 5 330 30303350 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002106.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          epimerase/racemase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          molecular_function
          catalytic activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          3-ketoglucose-reductase activity
          coenzyme binding
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000958 AK049709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000958 NM_001082975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000958 AI843700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000958 AK122247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000958 AK076856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149575 BC087941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149575 AK049709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149575 BC031783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149575 BC058857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149575 NM_001082975.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355538 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130353 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000958 AK049709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000958 NM_001082975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000958 AI843700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000958 AK122247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000958 AK076856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149575 BC087941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149575 AK049709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149575 BC031783.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149575 BC058857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149575 NM_001082975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206939 AK047709.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512541 BC087941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512541 AK049709.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512541 NM_001082975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512541 AK122247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512541 AK076856.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11586365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11586380 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11586368 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11586362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29620543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11143746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11680725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11532646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11826036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11469016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11670280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11586362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29620538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11586380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420224 - -
          Affymetrix HT MG-430 PM Array Plate 1428695_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459269_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420216 - -
          Mouse Genome 430 2.0 Array 1459269_PM_at - -
          Mouse Genome 430 2.0 Array 1428695_at - -
          Mouse Genome 430 2.0 Array 1459269_at - -
          Mouse Genome 430 2.0 Array 1428695_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 654795_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 654795_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400016227.mm.1 NM_001082975.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016227.mm.1 NM_001082975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016227.mm.1 TC1400002106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016228.mm.1 NM_001082975.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016228.mm.1 NM_001082975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016228.mm.1 TC1400002106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016230.mm.1 NM_001082975.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016230.mm.1 NM_001082975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016230.mm.1 TC1400002106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016229.mm.1 NM_001082975.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016229.mm.1 NM_001082975.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016229.mm.1 TC1400002106.mm.1 MTA TC
          Mouse Expression Set 430 1428695_at - -
          Mouse Expression Set 430 1428695_PM_at - -
          Mouse Expression Set 430 1459269_at - -
          Mouse Expression Set 430 1459269_PM_at - -
          Murine Genome U74v2 Set 133596_PM_at - -
          Murine Genome U74v2 Set 133596_at - -
          Affymetrix HT Mouse Genome 430B Array 1459269_at - -
          Affymetrix HT Mouse Genome 430B Array 1428695_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459269_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428695_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485236 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103990594 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215315 - -
          mouse-6 v1.1 (Illumina) scl000416.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 103990594 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103990594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2467578 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline