Search
Search

Naa30| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081430.1 | NP_001074899.1 | 3 - 4 | 1059 | 86 | |
| GenBank mRNA | AK015640.1 | - | 3 - 4 | 1032 | 86 | |
| AK044235.1 | - | 2 - 3 | 679 | 86 | ||
| BC054060.1 | - | 1 - 2 | 286 | 86 | ||
| BC157931.1 | - | 3 - 4 | 952 | 9088104 | 86 | |
| BC158065.1 | - | 3 - 4 | 952 | 9007364 | 86 | |
| BC172029.1 | - | 3 - 4 | 952 | 9054315 | 86 | |
Set Membership: |
|