Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01314699_m1
          See other RHOX13 GE Assays ›
          Gene Symbol:
          Rhox13
          Gene Name:
          reproductive homeobox 13
          Gene Aliases:
          1700123J19Rik
          Chromosome Location:
          Chr.X: 38120840 - 38129966 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01314699_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhox13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001185002.1 NP_001171931.1 2 - 3 1029 71
          GenBank mRNA AK007253.1 - 2 - 3 171 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000361.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to retinoic acid
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167715 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P474029 NM_001185002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P474029 AK007253.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P474029 NM_001185002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P474029 AK007253.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09267723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167715 - -
          Whole Mouse Genome G4122A A_51_P474029 NM_001185002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P474029 AK007253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P474029 NM_001185002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P474029 AK007253.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14745006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14814107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15186364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326408 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1443844_PM_at - -
          Mouse Genome 430 2.0 Array 1443844_at - -
          Mouse Genome 430 2.0 Array 1443844_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73614_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73614_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00002477.mm.1 NM_001185002.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002477.mm.1 NM_001185002.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002477.mm.1 TC0X00000361.mm.1 MTA TC
          Mouse Expression Set 430 1443844_PM_at - -
          Mouse Expression Set 430 1443844_at - -
          Murine Genome U74v2 Set 167327_at - -
          Murine Genome U74v2 Set 167327_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443844_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443844_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 738071 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237160 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline