Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01314921_g1
          See other RPS17 GE Assays ›
          Gene Symbol:
          Rps17
          Gene Name:
          ribosomal protein S17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 81342733 - 81345234 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01314921_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009092.3 NP_033118.1 2 - 3 182 83
          GenBank mRNA AK007992.1 - 2 - 3 190 83
          AK010647.1 - 2 - 3 184 83
          AK012781.1 - 2 - 3 171 83
          AK158902.1 - 1 - 2 248 83
          AK162022.1 - 2 - 3 176 83
          AK162335.1 - 2 - 3 174 83
          AK167206.1 - 2 - 3 176 83
          BC081466.1 - 1 - 2 169 6412006 83
          BC086900.1 - 2 - 3 175 6760363 83
          BC086901.1 - 2 - 3 175 6763239 83
          D25213.1 - 1 - 2 163 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Mus musculus Mouse Endogenous Controls 1700 Inventoried Mouse_Endogenous_Controls Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps17

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          rRNA processing
          translation
          erythrocyte homeostasis
          ribosomal small subunit biogenesis
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271555 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271554 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271557 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989125 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275309 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116793 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169721 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P482473 AK012781.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P482473 AK158902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P482473 AK162022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P482473 AK162335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P482473 AK010647.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25271570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116793 - -
          Whole Mouse Genome G4122A A_51_P482473 AK012781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P482473 AK158902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P482473 AK162022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P482473 AK162335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P482473 AK010647.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110205 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110979 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134293 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06325931 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271554 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271555 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25271556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25271514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06325940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06169129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06632984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06325929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06325934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06169128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06443773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25271524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06066204 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565086 - -
          Affymetrix HT MG-430 PM Array Plate 1455662_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1438986_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1437510_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10604597 - -
          Affymetrix Mouse Gene 1.1 ST Array 10374476 - -
          Affymetrix Mouse Gene 1.1 ST Array 10565083 - -
          Mouse Genome 430 2.0 Array 1438986_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438776_PM_x_at - -
          Mouse Genome 430 2.0 Array 1459986_PM_a_at - -
          Mouse Genome 430 2.0 Array 1459986_a_at - -
          Mouse Genome 430 2.0 Array 1437510_x_at - -
          Mouse Genome 430 2.0 Array 1437510_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438502_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438776_x_at - -
          Mouse Genome 430 2.0 Array 1438502_x_at - -
          Mouse Genome 430 2.0 Array 1438986_x_at - -
          Mouse Genome 430 2.0 Array 1455662_x_at - -
          Mouse Genome 430 2.0 Array 1455662_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0700030668.mm.1 NM_009092.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030668.mm.1 NM_009092.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030668.mm.1 TC0700003607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700030667.mm.1 NM_009092.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030667.mm.1 NM_009092.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030667.mm.1 TC0700003607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700030665.mm.1 NM_009092.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030665.mm.1 NM_009092.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030665.mm.1 TC0700003607.mm.1 MTA TC
          Mouse Expression Set 430 1459986_a_at - -
          Mouse Expression Set 430 1455662_x_at - -
          Mouse Expression Set 430 1438986_PM_x_at - -
          Mouse Expression Set 430 1438986_x_at - -
          Mouse Expression Set 430 1438501_PM_at - -
          Mouse Expression Set 430 1455662_PM_x_at - -
          Mouse Expression Set 430 1438776_x_at - -
          Mouse Expression Set 430 1437510_x_at - -
          Mouse Expression Set 430 1438501_at - -
          Mouse Expression Set 430 1437510_PM_x_at - -
          Mouse Expression Set 430 1459986_PM_a_at - -
          Mouse Expression Set 430 1438502_PM_x_at - -
          Mouse Expression Set 430 1438502_x_at - -
          Mouse Expression Set 430 1438776_PM_x_at - -
          Murine Genome U74v2 Set 102129_PM_at - -
          Murine Genome U74v2 Set 99590_PM_at - -
          Murine Genome U74v2 Set 99590_at - -
          Murine Genome U74v2 Set 102129_at - -
          Mouse Genome 430A 2.0 Array 1455662_x_at - -
          Mouse Genome 430A 2.0 Array 1455662_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437510_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438502_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438502_x_at - -
          Mouse Genome 430A 2.0 Array 1438776_x_at - -
          Mouse Genome 430A 2.0 Array 1438501_PM_at - -
          Mouse Genome 430A 2.0 Array 1438776_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438501_at - -
          Mouse Genome 430A 2.0 Array 1438986_x_at - -
          Mouse Genome 430A 2.0 Array 1459986_a_at - -
          Mouse Genome 430A 2.0 Array 1459986_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1437510_x_at - -
          Mouse Genome 430A 2.0 Array 1438986_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437510_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455662_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437510_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438986_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455662_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438986_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900479 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2360528 - -
          Illumina mouse-6 v1.0 expression beadchip 2360528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675747 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675747 - -
          mouse-6 v1.1 (Illumina) scl020068.3_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020068.3_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675747 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675747 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020068.3_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline