Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315040_s1
          See other ZMYM5 GE Assays ›
          Gene Symbol:
          Zmym5
          Gene Name:
          zinc finger, MYM-type 5
          Gene Aliases:
          9830124H08Rik
          Chromosome Location:
          Chr.14: 56790585 - 56811896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315040_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zmym5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253752.1 NP_001240681.1 8 3284 85
          NM_001253753.1 NP_001240682.1 8 3354 85
          NM_144842.3 NP_659091.3 8 3348 85
          GenBank mRNA AK165863.1 - 1 1208 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zmym5

          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624840 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041050 AK165863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041050 NM_144842.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2463930 AK155333.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848325 AK085200.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848325 AK085233.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848325 XM_001480114.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848325 XM_001480324.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160113 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113440 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166794 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2463930 AK155333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041050 AK165863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041050 NM_144842.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29624854 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166794 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08121931 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08121967 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113440 - -
          Whole Mouse Genome G4122A A_52_P288288 CN534727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288288 BC027380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288288 AK156501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288288 AK036513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288288 AK155333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P179211 BC058644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225981 AK165863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225981 BC027380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225981 AK156501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225981 AK163025.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225981 AK165292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277695 AK165863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277695 AK156501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277695 AK163025.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277695 AK165292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277695 NM_144842.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106027 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166794 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11293674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624840 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29624841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29624775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11480105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11480101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11372645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11482898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11482896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11200585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11368077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11556357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29624838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420354 - -
          Affymetrix HT MG-430 PM Array Plate 1425577_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442007_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445543_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420348 - -
          Mouse Genome 430 2.0 Array 1442007_at - -
          Mouse Genome 430 2.0 Array 1425577_PM_at - -
          Mouse Genome 430 2.0 Array 1442007_PM_at - -
          Mouse Genome 430 2.0 Array 1445543_PM_at - -
          Mouse Genome 430 2.0 Array 1445543_at - -
          Mouse Genome 430 2.0 Array 1425577_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219105_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219105_at - -
          Affymetrix Mouse Clariom S Array PSR1400016438.mm.1 NM_001253752.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016438.mm.1 NM_001253752.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016438.mm.1 TC1400002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 NM_001253752.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 NM_001253752.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 NM_001253753.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 NM_001253753.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 NM_144842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016427.mm.1 TC1400002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016443.mm.1 NM_001253752.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016443.mm.1 NM_001253752.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016443.mm.1 NM_144842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016443.mm.1 TC1400002132.mm.1 MTA TC
          Mouse Expression Set 430 1425577_at - -
          Mouse Expression Set 430 1425577_PM_at - -
          Mouse Expression Set 430 1445543_PM_at - -
          Mouse Expression Set 430 1442007_PM_at - -
          Mouse Expression Set 430 1445543_at - -
          Mouse Expression Set 430 1442007_at - -
          Murine Genome U74v2 Set 135351_i_at - -
          Murine Genome U74v2 Set 135351_PM_i_at - -
          Murine Genome U74v2 Set 166543_PM_r_at - -
          Murine Genome U74v2 Set 135352_r_at - -
          Murine Genome U74v2 Set 135352_PM_r_at - -
          Murine Genome U74v2 Set 166543_r_at - -
          Mouse Genome 430A 2.0 Array 1425577_at - -
          Mouse Genome 430A 2.0 Array 1425577_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425577_at - -
          Affymetrix HT Mouse Genome 430A Array 1425577_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442007_at - -
          Affymetrix HT Mouse Genome 430B Array 1445543_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442007_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445543_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 762461 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6130072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231971 - -
          mouse-6 v1.1 (Illumina) scl0219105.1_166-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0219105.1_166-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231971 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0219105.1_166-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-g7ph8:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0