Search
Search

Wdr73| Gene Symbol: Wdr73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028026.2 | NP_082302.2 | 3 - 4 | 240 | 70 | |
| NR_027976.1 | 2 - 3 | 713 | 70 | |||
| GenBank mRNA | AK004708.1 | - | 2 - 3 | 678 | 70 | |
| AK010448.1 | - | 3 - 4 | 206 | 70 | ||
| AK034740.1 | - | 3 - 4 | 204 | 70 | ||
| AK049132.1 | - | 3 - 4 | 238 | 70 | ||
| AK169377.1 | - | 2 - 3 | 677 | 70 | ||
| AK169527.1 | - | 3 - 4 | 240 | 70 | ||
| AK220207.1 | - | 3 - 4 | 209 | 70 | ||
| BC028870.1 | - | 3 - 4 | 193 | 4037614 | 70 | |
| BC068122.1 | - | 2 - 3 | 667 | 70 | ||
| BC087940.1 | - | 3 - 4 | 199 | 30302907 | 70 | |
Set Membership: |
|