Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315446_m1
          See other MARC1 GE Assays ›
          Gene Symbol:
          Marc1
          Gene Name:
          mitochondrial amidoxime reducing component 1
          Gene Aliases:
          1300013F15Rik, Mosc1
          Chromosome Location:
          Chr.1: 184786767 - 184811308 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Marc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290273.1 NP_001277202.1 2 - 3 628 67
          GenBank mRNA AK004989.1 - 2 - 3 525 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nitrate metabolic process
          oxidation-reduction process
          catalytic activity
          N-ethylmaleimide reductase activity
          nitrate reductase activity
          oxidoreductase activity
          molybdenum ion binding
          pyridoxal phosphate binding
          molybdopterin cofactor binding
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072453 AK011722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072453 BC028441.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072453 NM_001081361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072453 AK004989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072611 NM_001081361.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829714 BC055771.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829714 NM_001031664.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127961 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113250 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161327 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072453 AK011722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072453 BC028441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072453 NM_001081361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072453 AK004989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072611 NM_001081361.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113250 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00271794 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161327 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00271798 - -
          Whole Mouse Genome G4122A A_51_P454696 AK011722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454696 BC028441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454696 NM_001081361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454696 AK004989.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161327 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20897408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00175375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00170218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00540204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00237576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00501016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00156531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00787262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00237574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00540203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00540206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00501015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00892697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00360154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00604483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00175379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00536241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00921630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00122697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20897436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360847 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360845 - -
          Affymetrix Mouse Gene 1.1 ST Array 10360840 - -
          Mouse Genome 430 2.0 Array 1446074_PM_at - -
          Mouse Genome 430 2.0 Array 1446074_at - -
          Mouse Genome 430 2.0 Array 1428005_at - -
          Mouse Genome 430 2.0 Array 1428005_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66112_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66112_at - -
          Mouse Expression Set 430 1428005_at - -
          Mouse Expression Set 430 1428005_PM_at - -
          Mouse Expression Set 430 1446074_at - -
          Mouse Expression Set 430 1446074_PM_at - -
          Mouse Genome 430A 2.0 Array 1428005_PM_at - -
          Mouse Genome 430A 2.0 Array 1428005_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 730357 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2752016 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2752016 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline