Search
Search

Gnrh1| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008145.2 | NP_032171.1 | 2 - 3 | 240 | 97 | |
| GenBank mRNA | AK139480.1 | - | 2 - 3 | 202 | 97 | |
| BC116897.1 | - | 1 - 2 | 212 | 40126216 | 97 | |
| BC116899.1 | - | 1 - 2 | 212 | 40126218 | 97 | |
| CJ059442.1 | - | 2 - 3 | 202 | 97 | ||
| CX763652.1 | - | 2 - 3 | 240 | 97 | ||
Set Membership: |
|