Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315850_m1
          See other GM5149 GE Assays ›
          Gene Symbol:
          Gm5149
          Gene Name:
          predicted gene 5149
          Gene Aliases:
          EG381483
          Chromosome Location:
          Chr.3: 149335731 - 149340637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315850_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5149
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK132813.1 - 201 86

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          primary active transporter

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05116325 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05116344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00788036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05116374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05116360 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05116374 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02906446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02700019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02295246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02618777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03042927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03114498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02375208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02441436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02845008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02685018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02555299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02481796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03047385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03003740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02700026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02631150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02434726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02906441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02481797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02700006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03152338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02685007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02734967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02555287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03037284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03152339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02991882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02646740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22632709 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 688526 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2528962 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2814524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2814524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline