Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315945_s1
          See other OLFR728 GE Assays ›
          Gene Symbol:
          Olfr728
          Gene Name:
          olfactory receptor 728
          Gene Aliases:
          MOR246-1P
          Chromosome Location:
          Chr.14: 50139702 - 50140637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315945_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr728
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011809.1 NP_001011809.1 1 893 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr728

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184831 NM_001011809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174243 NM_001011809.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646922 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184831 NM_001011809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174243 NM_001011809.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596494 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08108403 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08108403 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11791851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11791848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11384771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11212283 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11212274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11384750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11212279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419441 - -
          Affymetrix Mouse Gene 1.1 ST Array 10419440 - -
          Affymetrix Mouse Clariom S Array PSR1400014828.mm.1 NM_001011809.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014828.mm.1 NM_001011809.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014828.mm.1 TC1400001975.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 748746 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160356 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160356 - -
          mouse-6 v1.1 (Illumina) GI_58801383-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160356 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160356 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline