Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315966_s1
          See other OLFR744 GE Assays ›
          Gene Symbol:
          Olfr744
          Gene Name:
          olfactory receptor 744
          Gene Aliases:
          MOR106-13P
          Chromosome Location:
          Chr.14: 50618224 - 50619195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315966_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr744
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011738.1 NP_001011738.1 1 929 108
          GenBank mRNA BC138726.1 - 1 992 8861748 108
          BC138727.1 - 1 992 8861749 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr744

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134396 NM_001011738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134396 BC138726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134396 BC138727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134396 NM_001011738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134396 BC138726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134396 BC138727.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199447 NM_001011738.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199447 AF121973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199447 BC138726.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199447 BC138727.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597857 - -
          Whole Mouse Genome G4122A A_51_P199447 NM_001011738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199447 AF121973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199447 BC138726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199447 BC138727.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414488 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414487 - -
          Affymetrix Mouse Clariom S Array PSR1400004433.mm.1 NM_001011738.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004433.mm.1 NM_001011738.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004433.mm.1 TC1400000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004432.mm.1 TC1400000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004434.mm.1 TC1400000561.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 341366 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161447 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161447 - -
          mouse-6 v1.1 (Illumina) GI_58801269-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161447 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline