Search
Search

Fgfr1op| Gene Symbol: Fgfr1op | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197046.1 | NP_001183975.1 | 5 - 6 | 522 | 93 | |
| NM_201230.5 | NP_957682.1 | 5 - 6 | 522 | 93 | ||
| GenBank mRNA | AK016110.1 | - | 5 - 6 | 525 | 93 | |
| AK028778.1 | - | 5 - 6 | 451 | 93 | ||
| AK029470.1 | - | 5 - 6 | 521 | 93 | ||
| AK031261.1 | - | 5 - 6 | 505 | 93 | ||
| AK050389.1 | - | 5 - 6 | 460 | 93 | ||
| BC050774.1 | - | 5 - 6 | 474 | 93 | ||
| BC058804.1 | - | 5 - 6 | 457 | 93 | ||
| BC080717.1 | - | 5 - 6 | 509 | 4935625 | 93 | |
| BC108331.1 | - | 5 - 6 | 520 | 30434232 | 93 | |
Set Membership: |
|