Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316525_m1
          See other 4921511H03RIK GE Assays ›
          Gene Symbol:
          4921511H03Rik
          Gene Name:
          RIKEN cDNA 4921511H03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 7302505 - 7311491 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316525_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4921511H03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027603.2 NP_081879.1 2 - 3 880 79
          GenBank mRNA AK076678.1 - 2 - 3 881 79
          BC049718.1 - 2 - 3 917 6742354 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000040.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4921511H03Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23463652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23463654 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK133311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 BC049718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 NM_027603.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK014870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK076678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK133311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 BC049718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 NM_027603.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK014870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161052 AK076678.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148468 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK133311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 BC049718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 NM_027603.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK014870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK076678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK133311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 BC049718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 NM_027603.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK014870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161052 AK076678.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23463394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148468 - -
          Whole Mouse Genome G4122A A_51_P147906 AK133311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147906 AK014870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK133311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 BC049718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 NM_027603.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK014870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK076678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK133311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 BC049718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 NM_027603.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK014870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161052 AK076678.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121588 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04427482 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247946 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23463652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23463654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23463452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04427484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04128005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04247944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04247947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04166347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04231621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04766536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04766538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23463418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519493 - -
          Affymetrix HT MG-430 PM Array Plate 1430467_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519490 - -
          Mouse Genome 430 2.0 Array 1430467_PM_at - -
          Mouse Genome 430 2.0 Array 1430467_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70920_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70920_at - -
          Affymetrix Mouse Clariom S Array PSR0500000345.mm.1 TC0500000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000345.mm.1 NM_027603.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000345.mm.1 NM_027603.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000340.mm.1 TC0500000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000340.mm.1 NM_027603.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000340.mm.1 NM_027603.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000342.mm.1 TC0500000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000342.mm.1 NM_027603.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000342.mm.1 NM_027603.2 Refseq
          Mouse Expression Set 430 1430467_PM_at - -
          Mouse Expression Set 430 1430467_at - -
          Affymetrix HT Mouse Genome 430B Array 1430467_at - -
          Affymetrix HT Mouse Genome 430B Array 1430467_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4780167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780167 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705700 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705700 - -
          mouse-6 v1.1 (Illumina) scl26902.2.300_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26902.2.300_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2705700 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2911237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2911237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705700 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0