Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01317809_m1
          See other 2610034B18RIK GE Assays ›
          Gene Symbol:
          2610034B18Rik
          Gene Name:
          RIKEN cDNA 2610034B18 gene
          Gene Aliases:
          Arpin
          Chromosome Location:
          Chr.7: 79925359 - 79935264 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01317809_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610034B18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027420.4 NP_081696.1 5 - 6 692 98
          GenBank mRNA AK011654.1 - 5 - 6 750 98
          AK039350.1 - 5 - 6 700 98
          AK051261.1 - 5 - 6 693 98
          AK156013.1 - 5 - 6 760 98
          BC031379.1 - 5 - 6 691 5360750 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003574.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2610034B18Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032498 AK156013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032498 AK011654.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032498 AK051261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032498 NM_027420.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164754 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145488 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032498 AK156013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032498 AK011654.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032498 AK051261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032498 NM_027420.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25264024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25263993 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01557882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145488 - -
          Whole Mouse Genome G4122A A_52_P615010 AK156013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615010 BC031379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615010 AK039350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615010 AK051261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615010 NM_027420.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381157 AK051261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381157 NM_027420.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103544 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06214129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06214125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06109488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06109491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06109490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06214124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06109485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25264017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25263931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06052231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05962555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06660201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06512063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06294276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05962562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05967454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06660204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06512064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06109485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06479588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06451409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25263951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25264009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10564853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564851 - -
          Affymetrix HT MG-430 PM Array Plate 1428450_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10564849 - -
          Mouse Genome 430 2.0 Array 1428450_PM_at - -
          Mouse Genome 430 2.0 Array 1428450_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70420_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70420_at - -
          Affymetrix Mouse Clariom S Array PSR0700030208.mm.1 NM_027420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030208.mm.1 NM_027420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030208.mm.1 TC0700003574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700030207.mm.1 NM_027420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030207.mm.1 NM_027420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030207.mm.1 TC0700003574.mm.1 MTA TC
          Mouse Expression Set 430 1428450_PM_at - -
          Mouse Expression Set 430 1428450_at - -
          Murine Genome U74v2 Set 135677_PM_f_at - -
          Murine Genome U74v2 Set 135677_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1428450_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428450_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617502 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754257 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754257 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline