Search
Search

1700006A11Rik| Gene Symbol: 1700006A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027939.1 | NP_082215.1 | 9 - 10 | 1420 | 123 | |
| GenBank mRNA | AK005660.1 | - | 8 - 9 | 1205 | 123 | |
| AK132965.1 | - | 9 - 10 | 1432 | 123 | ||
| BC137977.1 | - | 9 - 10 | 1420 | 8860996 | 123 | |
Set Membership: |
|