Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145514.5 | NP_663489.4 | 3 - 4 | 1105 | 91 | |
GenBank mRNA | AK075743.1 | - | 3 - 4 | 497 | 91 | |
AK082326.1 | - | 4 - 5 | 400 | 91 | ||
AK184747.1 | - | 3 - 4 | 270 | 91 | ||
AK205717.1 | - | 2 - 3 | 190 | 91 | ||
BC058601.1 | - | 1 - 2 | 101 | 91 | ||
BC139336.1 | - | 3 - 4 | 267 | 8862358 | 91 | |
BC139337.1 | - | 3 - 4 | 267 | 8862359 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |