Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318436_g1
          See other PGPEP1L GE Assays ›
          Gene Symbol:
          Pgpep1l
          Gene Name:
          pyroglutamyl-peptidase I-like
          Gene Aliases:
          C330024D12Rik
          Chromosome Location:
          Chr.7: 68236450 - 68264484 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318436_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgpep1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030101.1 NP_084377.1 3 - 4 439 70
          GenBank mRNA AK021221.1 - 3 - 4 440 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003458.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Protease 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          pyroglutamyl-peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174323 NM_030101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174323 AK021221.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126604 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148316 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174323 NM_030101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174323 AK021221.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06228286 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01538428 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148316 - -
          Whole Mouse Genome G4122A A_51_P316243 NM_030101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P316243 AK021221.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126604 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06170088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06620046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06620040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06369442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25198946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430210_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10564502 - -
          Mouse Genome 430 2.0 Array 1430210_at - -
          Mouse Genome 430 2.0 Array 1430210_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78444_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78444_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700029452.mm.1 NM_030101.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029452.mm.1 NM_030101.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029452.mm.1 TC0700003458.mm.1 MTA TC
          Mouse Expression Set 430 1430210_at - -
          Mouse Expression Set 430 1430210_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430210_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430210_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 776901 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714212 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714212 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline