Search
Search

Fam169b| Gene Symbol: Fam169b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013811.2 | NP_001013833.1 | 4 - 5 | 369 | 130 | |
| GenBank mRNA | BC039209.1 | - | 4 - 5 | 395 | 4161789 | 130 |
| BC039945.1 | - | 4 - 5 | 406 | 4161789 | 130 | |
Set Membership: |
|