Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318496_g1
          See other TMEM126A GE Assays ›
          Gene Symbol:
          Tmem126a
          Gene Name:
          transmembrane protein 126A
          Gene Aliases:
          1810020E01Rik
          Chromosome Location:
          Chr.7: 90450712 - 90457208 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318496_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem126a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025460.2 NP_079736.1 4 - 5 537 84
          GenBank mRNA AK007559.1 - 4 - 5 538 84
          AK050267.1 - 4 - 5 508 84
          AK210038.1 - 2 - 3 214 84
          BC023171.1 - 4 - 5 501 4946998 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem126a

          Gene Ontology Categories:

          Function(s) Process(es)

          optic nerve development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313745 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313746 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313742 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 NM_025460.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK005343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK007559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK050267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 BC023171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 NM_025460.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK005343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK007559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 AK050267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233815 BC023171.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78670 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127070 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 NM_025460.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK005343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK007559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK050267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 BC023171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 NM_025460.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK005343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK007559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 AK050267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233815 BC023171.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25313756 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127070 - -
          Whole Mouse Genome G4122A A_51_P233815 NM_025460.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK005343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK007559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK050267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 BC023171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 NM_025460.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK005343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK007559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 AK050267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233815 BC023171.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127070 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06488211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06280696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06020912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06280698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313745 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313742 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25313746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25313638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06622481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06146630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06426933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06018224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06622460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06018226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06488210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05923453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565515 - -
          Affymetrix HT MG-430 PM Array Plate 1451000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10565514 - -
          Mouse Genome 430 2.0 Array 1451000_at - -
          Mouse Genome 430 2.0 Array 1451000_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66271_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66271_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700031434.mm.1 NM_025460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031434.mm.1 NM_025460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031434.mm.1 TC0700003726.mm.1 MTA TC
          Mouse Expression Set 430 1451000_PM_at - -
          Mouse Expression Set 430 1451000_at - -
          Murine Genome U74v2 Set 94508_at - -
          Murine Genome U74v2 Set 94508_PM_at - -
          Mouse Genome 430A 2.0 Array 1451000_at - -
          Mouse Genome 430A 2.0 Array 1451000_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451000_at - -
          Affymetrix HT Mouse Genome 430A Array 1451000_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5130121 - -
          Illumina mouse-6 v1.0 expression beadchip 5130121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659774 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659774 - -
          mouse-6 v1.1 (Illumina) scl31053.2.1_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31053.2.1_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803627 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803627 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31053.2.1_38-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline