Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318783_m1
          See other CIB1 GE Assays ›
          Gene Symbol:
          Cib1
          Gene Name:
          calcium and integrin binding 1 (calmyrin)
          Gene Aliases:
          Cibkip, Kip, Prkdcip
          Chromosome Location:
          Chr.7: 80227156 - 80232805 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318783_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cib1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291275.1 NP_001278204.1 6 - 7 786 97
          NM_001291276.1 NP_001278205.1 4 - 5 657 97
          NM_011870.5 NP_036000.1 6 - 7 801 97
          GenBank mRNA AB006463.1 - 6 - 7 582 97
          AF173010.1 - 6 - 7 580 97
          AK010345.1 - 6 - 7 598 97
          AK027915.1 - 6 - 7 613 97
          AK088450.1 - 4 - 5 472 97
          AK152805.1 - 6 - 7 633 97
          AK165472.1 - 6 - 7 586 97
          BC003714.1 - 6 - 7 587 3599986 97
          BC054385.1 - 6 - 7 606 6476137 97

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cib1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          positive regulation of cell-matrix adhesion
          response to ischemia
          negative regulation of protein kinase activity
          apoptotic process
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle
          endomitotic cell cycle
          cell adhesion
          spermatogenesis
          spermatid development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          negative regulation of neuron projection development
          cell differentiation
          platelet formation
          positive regulation of cell growth
          positive regulation of cell migration
          cytoplasmic microtubule organization
          positive regulation of cell adhesion mediated by integrin
          thrombopoietin-mediated signaling pathway
          regulation of cell proliferation
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          negative regulation of megakaryocyte differentiation
          positive regulation of NF-kappaB transcription factor activity
          cell division
          regulation of cell division
          negative regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of calcineurin-NFAT signaling cascade
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          negative regulation of protein serine/threonine kinase activity
          positive regulation of protein serine/threonine kinase activity
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of protein targeting to membrane
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to nerve growth factor stimulus
          positive regulation of male germ cell proliferation
          calcium ion binding
          protein binding
          calcium-dependent protein kinase inhibitor activity
          Ras GTPase binding
          protein serine/threonine kinase inhibitor activity
          protein anchor
          ion channel binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265477 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265479 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982683 BC003714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982683 BC054385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982683 AK152805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982683 NM_011870.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982683 AK010345.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969893 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155724 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982683 BC003714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982683 BC054385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982683 AK152805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982683 NM_011870.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982683 AK010345.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123384 AK165472.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123384 BC003714.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123384 BC054385.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123384 AK152805.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123384 NM_011870.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175454 AK165472.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175454 BC003714.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175454 BC054385.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175454 AK152805.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175454 NM_011870.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265465 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06255807 - -
          Whole Mouse Genome G4122A A_51_P175454 AK165472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175454 BC003714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175454 BC054385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175454 AK152805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175454 NM_011870.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123384 AK165472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123384 BC003714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123384 BC054385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123384 AK152805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123384 NM_011870.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100876 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106225 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237995 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265479 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265477 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06417267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06237973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06101192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06438695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06361099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10564876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564872 - -
          Affymetrix HT MG-430 PM Array Plate 1448564_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425856_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10564869 - -
          Mouse Genome 430 2.0 Array 1425856_at - -
          Mouse Genome 430 2.0 Array 1448564_at - -
          Mouse Genome 430 2.0 Array 1425856_PM_at - -
          Mouse Genome 430 2.0 Array 1448564_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23991_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23991_PM_at - -
          Mouse Expression Set 430 1448564_PM_at - -
          Mouse Expression Set 430 1448564_at - -
          Mouse Expression Set 430 1425856_at - -
          Mouse Expression Set 430 1425856_PM_at - -
          Murine Genome U74v2 Set 96958_PM_at - -
          Murine Genome U74v2 Set 96958_at - -
          Murine Genome U74v2 Set 96957_PM_at - -
          Murine Genome U74v2 Set 96957_at - -
          Mouse Genome 430A 2.0 Array 1448564_at - -
          Mouse Genome 430A 2.0 Array 1448564_PM_at - -
          Mouse Genome 430A 2.0 Array 1425856_PM_at - -
          Mouse Genome 430A 2.0 Array 1425856_at - -
          Affymetrix HT Mouse Genome 430A Array 1425856_at - -
          Affymetrix HT Mouse Genome 430A Array 1425856_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448564_at - -
          Affymetrix HT Mouse Genome 430A Array 1448564_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621844 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621844 - -
          mouse-6 v1.1 (Illumina) scl31132.7.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31132.7.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621847 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621847 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31132.7.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline