Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01319122_m1
          See other NEDD8 GE Assays ›
          Gene Symbol:
          Nedd8
          Gene Name:
          neural precursor cell expressed, developmentally down-regulated gene 8
          Gene Aliases:
          Rub1
          Chromosome Location:
          Chr.14: 55662267 - 55671906 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01319122_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nedd8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008683.3 NP_032709.1 3 - 4 237 64
          GenBank mRNA AK146732.1 - 3 - 4 238 64
          AK147080.1 - 3 - 4 222 64
          AK158993.1 - 2 - 3 104 64
          BC004625.1 - 3 - 4 222 3585834 64
          D10918.1 - 3 - 4 204 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002094.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nedd8

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          protein localization
          response to organic cyclic compound
          protein neddylation
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619205 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 NM_008683.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK146732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK147080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK158993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 NM_008683.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK146732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK147080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 AK158993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 AF288525.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 AK005029.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 AK079323.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 AK137089.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 BC003458.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508827 NM_023627.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108772 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124997 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 NM_008683.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK146732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK147080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK158993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 BC004625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 NM_008683.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK146732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK147080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 AK158993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 NM_008683.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK146732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK147080.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK158993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 NM_008683.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK146732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK147080.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 AK158993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29619204 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124997 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08119182 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08119191 - -
          Whole Mouse Genome G4122A A_51_P416869 NM_008683.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK146732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK147080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK158993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 BC004625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 NM_008683.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK146732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK147080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 AK158993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416869 BC004625.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115680 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105805 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29619205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11255716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11383799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11274254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11307863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11784139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11560433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29619175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420092 - -
          Affymetrix HT MG-430 PM Array Plate 1423715_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420089 - -
          Mouse Genome 430 2.0 Array 1445811_PM_at - -
          Mouse Genome 430 2.0 Array 1423715_a_at - -
          Mouse Genome 430 2.0 Array 1423715_PM_a_at - -
          Mouse Genome 430 2.0 Array 1440996_PM_at - -
          Mouse Genome 430 2.0 Array 1445811_at - -
          Mouse Genome 430 2.0 Array 1440996_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18002_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18002_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400016067.mm.1 NM_008683.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016067.mm.1 NM_008683.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016067.mm.1 TC1400002094.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016069.mm.1 NM_008683.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016069.mm.1 NM_008683.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016069.mm.1 TC1400002094.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016068.mm.1 NM_008683.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016068.mm.1 NM_008683.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016068.mm.1 TC1400002094.mm.1 MTA TC
          Mouse Expression Set 430 1423715_PM_a_at - -
          Mouse Expression Set 430 1423715_a_at - -
          Murine Genome U74v2 Set 93519_s_at - -
          Murine Genome U74v2 Set 93519_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1423715_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423715_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423715_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423715_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416812 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6180632 - -
          Illumina mouse-6 v1.0 expression beadchip 6180632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256971 - -
          mouse-6 v1.1 (Illumina) scl45531.6.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45531.6.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256971 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45531.6.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline