Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01319369_m1
          See other EML2 GE Assays ›
          Gene Symbol:
          Eml2
          Gene Name:
          echinoderm microtubule associated protein like 2
          Gene Aliases:
          1600029N02Rik
          Chromosome Location:
          Chr.7: 19176266 - 19206482 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01319369_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eml2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162996.1 NP_001156468.1 7 - 8 644 62
          NM_028153.1 NP_082429.1 7 - 8 701 62
          GenBank mRNA AK005562.1 - 7 - 8 667 62
          AK184916.1 - 3 - 4 257 62
          AK213701.1 - 2 - 3 158 62
          BC055476.1 - 7 - 8 644 4218133 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000290.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of microtubule nucleation
          negative regulation of microtubule polymerization
          receptor binding
          microtubule binding
          protein C-terminus binding
          tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991903 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991902 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991904 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989921 AK215580.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989921 BC055476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989921 NM_001162996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989921 NM_028153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989921 AK005562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071811 AK200420.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071811 AK200422.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071811 NM_001162996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071811 NM_028153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071811 AK005562.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490520 AK005874.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490520 AK077203.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490520 BC109355.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490520 NM_029294.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15981 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137493 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129862 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071811 AK200420.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071811 AK200422.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071811 NM_001162996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071811 NM_028153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071811 AK005562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989921 AK215580.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989921 BC055476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989921 NM_001162996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989921 NM_028153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989921 AK005562.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06134455 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06134429 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06134447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129862 - -
          Whole Mouse Genome G4122A A_51_P428013 AK215580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428013 BC055476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428013 NM_001162996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428013 NM_028153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428013 AK005562.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129862 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06134447 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991902 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991903 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24991904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24991999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550620 - -
          Affymetrix HT MG-430 PM Array Plate 1428173_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550605 - -
          Mouse Genome 430 2.0 Array 1428173_at - -
          Mouse Genome 430 2.0 Array 1428173_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72205_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72205_at - -
          Affymetrix Mouse Clariom S Array PSR0700002223.mm.1 TC0700000290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002223.mm.1 NM_001162996.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002223.mm.1 NM_001162996.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002223.mm.1 NM_028153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002223.mm.1 NM_028153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002222.mm.1 TC0700000290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002222.mm.1 NM_001162996.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002222.mm.1 NM_001162996.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002222.mm.1 NM_028153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002222.mm.1 NM_028153.1 Refseq
          Mouse Expression Set 430 1428173_PM_at - -
          Mouse Expression Set 430 1428173_at - -
          Affymetrix HT Mouse Genome 430B Array 1428173_at - -
          Affymetrix HT Mouse Genome 430B Array 1428173_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932452 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101990010 - -
          Illumina mouse-6 v1.0 expression beadchip 4060411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229138 - -
          mouse-6 v1.1 (Illumina) scl00037.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00037.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601281 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00037.1_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline