| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100013903.mm.1 |
NM_001034899.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013903.mm.1 |
NM_001034899.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013903.mm.1 |
TC0100001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013900.mm.1 |
NM_001034899.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013900.mm.1 |
NM_001034899.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013900.mm.1 |
TC0100001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013902.mm.1 |
NM_001034899.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013902.mm.1 |
NM_001034899.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013902.mm.1 |
TC0100001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013899.mm.1 |
NM_001034899.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013899.mm.1 |
NM_001034899.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013899.mm.1 |
TC0100001678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013901.mm.1 |
NM_001034899.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013901.mm.1 |
NM_001034899.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013901.mm.1 |
TC0100001678.mm.1 |
MTA TC |