Search
Search

Gm16233| Gene Symbol: Gm16233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054018.1 | - | 157 | 84 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride channel calcium activated 4B | Clca4b |
AI747448, Clca7 |
-15.0kb | Anti-sense | Chr. 3 - 144910921 - 144932529 on NCBI Build GRCm38 |
| Downstream | chloride channel calcium activated 4A | Clca4a |
9130020L07Rik, AI504701, Clca4, Clca6 |
4.4kb | Anti-sense | Chr. 3 - 144952486 - 144975045 on NCBI Build GRCm38 |
Set Membership: |
|