Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01320215_m1
          See other CEACAM20 GE Assays ›
          Gene Symbol:
          Ceacam20
          Gene Name:
          carcinoembryonic antigen-related cell adhesion molecule 20
          Gene Aliases:
          9130012D09Rik
          Chromosome Location:
          Chr.7: 19965352 - 19991113 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01320215_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ceacam20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027839.2 NP_082115.2 11 - 12 1757 94
          GenBank mRNA AK018613.1 - 11 - 12 1754 94
          AK033581.2 - 11 - 12 1758 94
          BC049361.1 - 11 - 12 1738 6476373 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000315.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ceacam20

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047718 AK033581.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047718 AV374496.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047718 BC049361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047718 NM_027839.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952842 AK033581.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952842 BC049361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952842 NM_027839.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952842 AK018613.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139690 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047718 AK033581.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047718 AV374496.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047718 BC049361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047718 NM_027839.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952842 AK033581.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952842 BC049361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952842 NM_027839.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952842 AK018613.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24996044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03806487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03806305 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06136620 - -
          Whole Mouse Genome G4122A A_51_P228604 AK033581.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228604 BC049361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228604 NM_027839.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228604 AK018613.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06136620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03806487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24996026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24996021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550741 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550744 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550740 - -
          Mouse Genome 430 2.0 Array 1430383_at - -
          Mouse Genome 430 2.0 Array 1430383_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71601_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71601_at - -
          Affymetrix Mouse Clariom S Array PSR0700002482.mm.1 TC0700000315.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002482.mm.1 NM_027839.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002482.mm.1 NM_027839.2 Refseq
          Mouse Expression Set 430 1430383_at - -
          Mouse Expression Set 430 1430383_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 505152 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100670736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233831 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260131 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233831 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260131 - -
          Illumina Sentrix mouse-6.v1 bead array 100670736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100670736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline