Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01320772_m1
          See other 1700016C15RIK GE Assays ›
          Gene Symbol:
          1700016C15Rik
          Gene Name:
          RIKEN cDNA 1700016C15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 177728122 - 177753306 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01320772_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700016C15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027077.2 NP_081353.1 4 - 5 496 71
          GenBank mRNA AK006014.1 - 4 - 5 497 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001675.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225493 AK006014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225493 NM_027077.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225493 AK006014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225493 NM_027077.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66125 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145999 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145020 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225493 AK006014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225493 NM_027077.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225493 AK006014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225493 NM_027077.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145999 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145020 - -
          Whole Mouse Genome G4122A A_51_P225493 AK006014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225493 NM_027077.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225493 AK006014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225493 NM_027077.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145999 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20861527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01130911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00784783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00830246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00599063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00857537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00999370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00375071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00243386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01036482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00857536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01084508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01036481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00737078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00999371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00644306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00237718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00644307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00784777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20861476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10352099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352102 - -
          Affymetrix HT MG-430 PM Array Plate 1430674_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10352097 - -
          Mouse Genome 430 2.0 Array 1430674_at - -
          Mouse Genome 430 2.0 Array 1430674_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69428_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69428_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100013865.mm.1 NM_027077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013865.mm.1 NM_027077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013865.mm.1 TC0100001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013864.mm.1 NM_027077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013864.mm.1 NM_027077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013864.mm.1 TC0100001675.mm.1 MTA TC
          Mouse Expression Set 430 1430674_PM_at - -
          Mouse Expression Set 430 1430674_at - -
          Murine Genome U74v2 Set 167939_PM_s_at - -
          Murine Genome U74v2 Set 167939_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430674_at - -
          Affymetrix HT Mouse Genome 430B Array 1430674_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 372595 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222931 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222931 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0