Search
Search

Vcam1| Gene Symbol: Vcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011693.3 | NP_035823.3 | 2 - 3 | 656 | 71 | |
| GenBank mRNA | AK016465.1 | - | 2 - 3 | 426 | 71 | |
| AK030195.1 | - | 2 - 3 | 426 | 71 | ||
| AK143401.1 | - | 2 - 3 | 426 | 71 | ||
| AK162954.1 | - | 2 - 3 | 426 | 71 | ||
| BC011159.1 | - | 2 - 3 | 398 | 4206476 | 71 | |
| BC029823.1 | - | 2 - 3 | 656 | 4196681 | 71 | |
| CT010373.1 | - | 2 - 3 | 335 | 71 | ||
| L08431.1 | - | 2 - 3 | 395 | 71 | ||
| M84487.1 | - | 2 - 3 | 392 | 71 | ||
| U12884.1 | - | 2 - 3 | 378 | 71 | ||
| X67783.1 | - | 2 - 3 | 350 | 71 | ||
Set Membership: |
|