Search
Search

Slc39a8| Gene Symbol: Slc39a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135149.1 | NP_001128621.1 | 2 - 3 | 596 | 75 | |
| NM_001135150.1 | NP_001128622.1 | 2 - 3 | 746 | 75 | ||
| NM_026228.5 | NP_080504.3 | 2 - 3 | 705 | 75 | ||
| GenBank mRNA | AK014895.1 | - | 2 - 3 | 709 | 75 | |
| AK016853.1 | - | 2 - 3 | 483 | 75 | ||
| AK028215.1 | - | 2 - 3 | 494 | 75 | ||
| AK085740.1 | - | 2 - 3 | 600 | 75 | ||
| AK089060.1 | - | 1 - 2 | 605 | 75 | ||
| BC006731.1 | - | 1 - 2 | 407 | 3155670 | 75 | |
Set Membership: |
|