Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01322192_g1
          See other FITM1 GE Assays ›
          Gene Symbol:
          Fitm1
          Gene Name:
          fat storage-inducing transmembrane protein 1
          Gene Aliases:
          1110028A07Rik, AV010547, Cg10671, Fit1
          Chromosome Location:
          Chr.14: 55575674 - 55576952 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01322192_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fitm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026808.1 NP_081084.1 1 - 2 280 72
          GenBank mRNA AB054000.1 - 1 - 2 280 72
          AK003939.1 - 1 - 2 169 72
          BC037188.1 - 1 - 2 301 72
          BC048823.1 - 1 - 2 78 72
          BC104408.1 - 1 - 2 280 40050546 72
          BC104409.2 - 1 - 2 280 40050547 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000783.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fitm1

          Gene Ontology Categories:

          Function(s)

          phospholipid biosynthetic process
          positive regulation of sequestering of triglyceride
          lipid storage
          lipid particle organization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC104408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC104409.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC037188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC048823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 NM_026808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC104408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC104409.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC037188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 BC048823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164630 NM_026808.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299101 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC104408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC104409.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC037188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC048823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 NM_026808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC104408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC104409.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC037188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 BC048823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164630 NM_026808.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618583 - -
          Whole Mouse Genome G4122A A_51_P164630 BC104408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC104409.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC037188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC048823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 NM_026808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC104408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC104409.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC037188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 BC048823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164630 NM_026808.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11338901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11174413 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11174406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11338898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11174412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11174403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11174410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11338902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11681104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11541044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415280 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415281 - -
          Affymetrix HT MG-430 PM Array Plate 1451488_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415279 - -
          Mouse Genome 430 2.0 Array 1451488_at - -
          Mouse Genome 430 2.0 Array 1451488_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68680_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68680_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400006260.mm.1 NM_026808.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006260.mm.1 NM_026808.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006260.mm.1 TC1400000783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006261.mm.1 NM_026808.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006261.mm.1 NM_026808.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006261.mm.1 TC1400000783.mm.1 MTA TC
          Mouse Expression Set 430 1451488_PM_at - -
          Mouse Expression Set 430 1451488_at - -
          Mouse Genome 430A 2.0 Array 1451488_PM_at - -
          Mouse Genome 430A 2.0 Array 1451488_at - -
          Affymetrix HT Mouse Genome 430A Array 1451488_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451488_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 748128 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243516 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243516 - -
          mouse-6 v1.1 (Illumina) scl46277.2.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46277.2.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243516 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0