| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400007108.mm.1 |
NM_001243857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007108.mm.1 |
NM_001243857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007108.mm.1 |
TC1400000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400007111.mm.1 |
NM_001243857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007111.mm.1 |
NM_001243857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007111.mm.1 |
TC1400000882.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400007106.mm.1 |
NM_001243857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007106.mm.1 |
NM_001243857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400007106.mm.1 |
TC1400000882.mm.1 |
MTA TC |