Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01322262_s1
          See other IRF2BP1 GE Assays ›
          Gene Symbol:
          Irf2bp1
          Gene Name:
          interferon regulatory factor 2 binding protein 1
          Gene Aliases:
          6330414O09Rik, C530033F24
          Chromosome Location:
          Chr.7: 19004065 - 19006763 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01322262_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irf2bp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178757.3 NP_848872.2 1 2432 119
          GenBank mRNA AK032010.1 - 1 2377 119
          AK087457.1 - 1 2445 119
          BC019164.1 - 1 2113 3962494 119
          BC036162.1 - 1 2432 4984359 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irf2bp1

          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription corepressor activity
          transcription factor binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990860 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990867 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990864 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 BC019164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 BC036162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK087457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK083013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK031834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 BC019164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 BC036162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK087457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK083013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P381953 AK031834.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885653 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136730 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 BC019164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 BC036162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK087457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK083013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK031834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 BC019164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 BC036162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK087457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK083013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P381953 AK031834.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24990853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136730 - -
          Whole Mouse Genome G4122A A_52_P381953 BC019164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 BC036162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK087457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK083013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK031834.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 BC019164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 BC036162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK087457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK083013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P381953 AK031834.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133973 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06424705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06424695 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990860 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990867 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24990862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24990863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06145800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05889297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06145806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05911818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05889307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05889290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24990892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550528 - -
          Affymetrix HT MG-430 PM Array Plate 1451252_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550527 - -
          Mouse Genome 430 2.0 Array 1451252_at - -
          Mouse Genome 430 2.0 Array 1451252_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 272359_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 272359_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002028.mm.1 TC0700000281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002028.mm.1 NM_178757.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002028.mm.1 NM_178757.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002029.mm.1 TC0700000281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002029.mm.1 NM_178757.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002029.mm.1 NM_178757.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002030.mm.1 TC0700000281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002030.mm.1 NM_178757.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002030.mm.1 NM_178757.3 Refseq
          Mouse Expression Set 430 1451252_PM_at - -
          Mouse Expression Set 430 1451252_at - -
          Murine Genome U74v2 Set 100007_PM_at - -
          Murine Genome U74v2 Set 100007_at - -
          Mouse Genome 430A 2.0 Array 1451252_PM_at - -
          Mouse Genome 430A 2.0 Array 1451252_at - -
          Affymetrix HT Mouse Genome 430A Array 1451252_at - -
          Affymetrix HT Mouse Genome 430A Array 1451252_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 909676 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228568 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228568 - -
          mouse-6 v1.1 (Illumina) scl33017.2_469-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33017.2_469-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228568 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228568 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33017.2_469-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0