Search
Search

Dlgap5| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 11 - 12 | 1644 | 70 | |
| GenBank mRNA | AB076696.1 | - | 11 - 12 | 1644 | 70 | |
| AK048470.1 | - | 11 - 12 | 1414 | 70 | ||
| AK085882.1 | - | 11 - 12 | 1425 | 70 | ||
| AK129032.1 | - | 11 - 12 | 1390 | 70 | ||
| AK135820.1 | - | 11 - 12 | 1451 | 70 | ||
| AK147955.1 | - | 11 - 12 | 1414 | 70 | ||
| BC027273.1 | - | 11 - 12 | 1365 | 70 | ||
| BC043924.1 | - | 3 - 4 | 270 | 70 | ||
| BC058087.1 | - | 11 - 12 | 1378 | 6816660 | 70 | |
Set Membership: |
|