Search
Search
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293635.1 | NP_001280564.1 | 38 - 39 | 3770 | 93 | |
| NM_001293636.1 | NP_001280565.1 | 38 - 39 | 3818 | 93 | ||
| NM_008477.2 | NP_032503.2 | 39 - 40 | 3812 | 93 | ||
| GenBank mRNA | AJ517365.1 | - | 14 - 15 | 1063 | 93 | |
| AJ517366.1 | - | 14 - 15 | 1064 | 93 | ||
| AJ517367.1 | - | 14 - 15 | 1064 | 93 | ||
| AJ517368.1 | - | 15 - 16 | 1150 | 93 | ||
| AJ517369.1 | - | 15 - 16 | 1148 | 93 | ||
| AJ517370.2 | - | 15 - 16 | 1136 | 93 | ||
| AJ517371.1 | - | 17 - 18 | 1288 | 93 | ||
| AJ517372.1 | - | 16 - 17 | 1205 | 93 | ||
| AJ517373.1 | - | 16 - 17 | 1235 | 93 | ||
| AJ517374.1 | - | 15 - 16 | 1151 | 93 | ||
| AJ517375.1 | - | 16 - 17 | 1220 | 93 | ||
| AJ517376.1 | - | 16 - 17 | 1220 | 93 | ||
| AJ517377.1 | - | 17 - 18 | 1307 | 93 | ||
| AJ517378.1 | - | 14 - 15 | 1063 | 93 | ||
| AJ517379.1 | - | 14 - 15 | 1064 | 93 | ||
| AJ517380.1 | - | 16 - 17 | 1223 | 93 | ||
| AJ517381.1 | - | 16 - 17 | 1223 | 93 | ||
| AJ517382.1 | - | 15 - 16 | 1116 | 93 | ||
| AJ517383.1 | - | 16 - 17 | 1220 | 93 | ||
| AJ517384.1 | - | 15 - 16 | 1136 | 93 | ||
| AK163789.1 | - | 1 - 2 | 763 | 93 | ||
| AK172869.1 | - | 13 - 14 | 1467 | 93 | ||
| BC066871.1 | - | 38 - 39 | 3725 | 93 | ||
| BC075697.1 | - | 38 - 39 | 3737 | 93 | ||
| CD363975.1 | - | 6 - 7 | 509 | 93 | ||
| DT913164.1 | - | 5 - 6 | 383 | 93 | ||
| L43326.1 | - | 39 - 40 | 3800 | 93 | ||
Set Membership: |
|