Search
Search
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008523.2 | NP_032549.2 | 7 - 8 | 975 | 79 | |
| NM_203345.2 | NP_976220.2 | 13 - 14 | 1821 | 79 | ||
| NM_206941.1 | NP_996824.1 | 8 - 9 | 1158 | 79 | ||
| NM_206942.2 | NP_996825.2 | 12 - 13 | 1638 | 79 | ||
| GenBank mRNA | BC047138.1 | - | 7 - 8 | 975 | 6389834 | 79 |
| BC141299.1 | - | 12 - 13 | 1614 | 9055911 | 79 | |
| M90470.1 | - | 13 - 14 | 1821 | 79 | ||
| X07984.1 | - | 7 - 8 | 949 | 79 | ||
| X52621.1 | - | 7 - 8 | 964 | 79 | ||
Set Membership: |
|