| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21216452 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21216454 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2017491 |
BC003333.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2017491 |
CX224390.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2017491 |
NM_001141982.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2017491 |
NM_030243.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2017491 |
AK002739.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828848 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
AK012440.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
AK090349.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
AK090391.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
AK154995.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
AK172353.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
BC036332.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
NM_153775.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428704 |
XM_001481316.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
16513 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702999 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153580 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
519015 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162011 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P152261 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P138481 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2017491 |
BC003333.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2017491 |
CX224390.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2017491 |
NM_001141982.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2017491 |
NM_030243.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2017491 |
AK002739.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216455 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216488 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216459 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216464 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216449 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216442 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216451 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216483 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216456 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216465 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216481 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216452 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216447 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216437 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216439 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216453 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216480 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216458 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216484 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216463 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216474 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216478 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216468 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216462 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216469 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216461 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216473 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216454 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216485 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216466 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216471 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216476 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216445 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21216443 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P138481 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P152261 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P166818 |
BC003333.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P166818 |
CX224390.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P166818 |
NM_001141982.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P166818 |
NM_030243.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P166818 |
AK002739.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P138481 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P152261 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01735427 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01735422 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01735426 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21216454 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21216452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216458 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216350 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216433 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216389 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216425 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216474 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216490 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216488 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216377 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216382 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216454 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216496 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216429 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216375 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216408 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216360 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216345 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216421 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216419 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216409 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216435 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216449 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216348 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216359 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216393 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216483 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216369 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216471 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216394 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216481 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216347 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216395 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216456 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216357 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216445 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216480 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216468 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216493 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216436 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216399 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216492 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216491 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216479 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216498 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216406 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216469 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216461 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216464 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216463 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21216400 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216454 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01735427 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216479 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216443 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01345771 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01234878 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216451 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216488 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216474 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01165453 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01345776 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216496 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01345772 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216481 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216490 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216444 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216453 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216480 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01335258 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216409 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216413 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216460 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216493 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216483 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01335253 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216463 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01311659 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216422 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216431 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01735422 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216425 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216419 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216421 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216415 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216343 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216408 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01152440 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02010448 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216484 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216435 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216424 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216497 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216498 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216429 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216447 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01735432 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216445 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01224234 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02203314 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216439 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216461 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216465 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216449 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01848646 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216417 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216492 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216471 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01418331 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216405 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216450 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216491 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216342 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216455 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216346 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216432 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216436 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216476 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216464 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216466 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216456 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21216442 |
- |
- |