Search
Search

F13a1| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166391.1 | NP_001159863.1 | 7 - 8 | 1045 | 77 | |
| NM_028784.3 | NP_083060.2 | 7 - 8 | 1070 | 77 | ||
| GenBank mRNA | AK030914.1 | - | 7 - 8 | 1045 | 77 | |
| AK036403.1 | - | 7 - 8 | 1071 | 77 | ||
| AK049092.1 | - | 7 - 8 | 1071 | 77 | ||
| AK163591.1 | - | 7 - 8 | 1045 | 77 | ||
| AK165311.1 | - | 7 - 8 | 1071 | 77 | ||
| BC040274.1 | - | 7 - 8 | 1061 | 3482233 | 77 | |
Set Membership: |
|