Search
Search

Alox8| Gene Symbol: Alox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009661.4 | NP_033791.1 | 5 - 6 | 710 | 57 | |
| GenBank mRNA | AK028724.1 | - | 5 - 6 | 706 | 57 | |
| AK158553.1 | - | 3 - 4 | 584 | 57 | ||
| BC015253.1 | - | 5 - 6 | 710 | 4221815 | 57 | |
| U93277.1 | - | 5 - 6 | 676 | 57 | ||
| Y14696.1 | - | 5 - 6 | 689 | 57 | ||
Set Membership: |
|