Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01325629_m1
          See other JAG2 GE Assays ›
          Gene Symbol:
          Jag2
          Gene Name:
          jagged 2
          Gene Aliases:
          D12Ggc2e, Serh, mJagged2-1, sm
          Chromosome Location:
          Chr.12: 112907803 - 112929764 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01325629_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jag2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010588.2 NP_034718.2 24 - 25 3107 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002443.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jag2

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          in utero embryonic development
          cell fate determination
          respiratory system process
          cell communication
          Notch signaling pathway
          multicellular organism development
          spermatogenesis
          sensory perception of sound
          auditory receptor cell fate commitment
          morphogenesis of embryonic epithelium
          cell differentiation
          regulation of cell adhesion
          T cell differentiation
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          gamma-delta T cell differentiation
          thymic T cell selection
          epithelial cell apoptotic process involved in palatal shelf morphogenesis
          Notch binding
          calcium ion binding
          protein binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 BC009082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 BC031134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 NM_010588.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 AF038572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 BC009082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 BC031134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 NM_010588.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451338 AF038572.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76357 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161491 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111294 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 BC009082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 BC031134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 NM_010588.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 AF038572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 BC009082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 BC031134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 NM_010588.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451338 AF038572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 BC009082.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 BC031134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 NM_010588.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 AF038572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 BC009082.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 BC031134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 NM_010588.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P451338 AF038572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 BC009082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 BC031134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 NM_010588.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 AF038572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 BC009082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 BC031134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 NM_010588.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451338 AF038572.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09751338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402828 - -
          Affymetrix HT MG-430 PM Array Plate 1426431_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426430_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402808 - -
          Mouse Genome 430 2.0 Array 1426430_at - -
          Mouse Genome 430 2.0 Array 1426431_at - -
          Mouse Genome 430 2.0 Array 1426430_PM_at - -
          Mouse Genome 430 2.0 Array 1426431_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16450_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16450_at - -
          Affymetrix Mouse Clariom S Array PSR1200016970.mm.1 NM_010588.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016970.mm.1 NM_010588.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016970.mm.1 TC1200002443.mm.1 MTA TC
          Mouse Expression Set 430 1426430_PM_at - -
          Mouse Expression Set 430 1426430_at - -
          Mouse Expression Set 430 1426431_PM_at - -
          Mouse Expression Set 430 1426431_at - -
          Murine Genome U74v2 Set 92510_at - -
          Murine Genome U74v2 Set 92510_PM_at - -
          Murine Genome U74v2 Set 165694_at - -
          Murine Genome U74v2 Set 165694_PM_at - -
          Mouse Genome 430A 2.0 Array 1426430_at - -
          Mouse Genome 430A 2.0 Array 1426431_at - -
          Mouse Genome 430A 2.0 Array 1426431_PM_at - -
          Mouse Genome 430A 2.0 Array 1426430_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426431_at - -
          Affymetrix HT Mouse Genome 430A Array 1426430_at - -
          Affymetrix HT Mouse Genome 430A Array 1426431_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426430_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 851162 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500341 - -
          Illumina mouse-6 v1.0 expression beadchip 1500341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626585 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2626585 - -
          mouse-6 v1.1 (Illumina) scl42013.23.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42013.23.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2626585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2626585 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline