Search
Search
| Gene Symbol: Pdcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026176.3 | NP_080452.2 | 2 - 3 | 368 | 56 | |
| GenBank mRNA | AK004704.1 | - | 2 - 3 | 1081 | 56 | |
| AK028668.1 | - | 2 - 3 | 258 | 56 | ||
| AK028814.1 | - | 2 - 3 | 258 | 56 | ||
| AK133747.1 | - | 2 - 3 | 278 | 56 | ||
| AK166983.1 | - | 2 - 3 | 258 | 56 | ||
| BC006578.1 | - | 2 - 3 | 268 | 3586612 | 56 | |
Set Membership: |
|