Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327445_g1
          See other ZDHHC12 GE Assays ›
          Gene Symbol:
          Zdhhc12
          Gene Name:
          zinc finger, DHHC domain containing 12
          Gene Aliases:
          1190004A01Rik
          Chromosome Location:
          Chr.2: 30090944 - 30093635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327445_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037762.1 NP_001032851.1 4 - 5 528 73
          NM_025428.2 NP_079704.2 4 - 5 570 73
          GenBank mRNA AK004475.1 - 4 - 5 471 73
          AK013074.1 - 4 - 5 558 73
          BC021432.1 - 4 - 5 510 5030263 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003262.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zdhhc12

          Gene Ontology Categories:

          Function(s) Process(es)

          protein palmitoylation
          zinc ion binding
          palmitoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          protein-cysteine S-palmitoyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101315 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101317 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 AK087676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 BC021432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 NM_001037762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 NM_025428.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 AK004475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 AK087676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 BC021432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 NM_001037762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 NM_025428.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153224 AK004475.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825306 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169348 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 AK087676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 BC021432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 NM_001037762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 NM_025428.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 AK004475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 AK087676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 BC021432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 NM_001037762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 NM_025428.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153224 AK004475.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169348 - -
          Whole Mouse Genome G4122A A_51_P153224 AK087676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 BC021432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 NM_001037762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 NM_025428.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 AK004475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 AK087676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 BC021432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 NM_001037762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 NM_025428.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153224 AK004475.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120716 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169348 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01884539 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02257347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01884561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01884542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101315 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01884542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481396 - -
          Affymetrix HT MG-430 PM Array Plate 1448857_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481393 - -
          Mouse Genome 430 2.0 Array 1448857_a_at - -
          Mouse Genome 430 2.0 Array 1448857_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66220_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66220_at - -
          Affymetrix Mouse Clariom S Array PSR0200027487.mm.1 NM_001037762.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027487.mm.1 NM_001037762.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027487.mm.1 NM_025428.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027487.mm.1 NM_025428.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027487.mm.1 TC0200003262.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027488.mm.1 NM_001037762.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027488.mm.1 NM_001037762.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027488.mm.1 NM_025428.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027488.mm.1 NM_025428.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027488.mm.1 TC0200003262.mm.1 MTA TC
          Mouse Expression Set 430 1448857_a_at - -
          Mouse Expression Set 430 1448857_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1448857_a_at - -
          Mouse Genome 430A 2.0 Array 1448857_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448857_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448857_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801933 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3450377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218402 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218402 - -
          mouse-6 v1.1 (Illumina) scl19480.4_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19480.4_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2520032 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3148355 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2520032 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3148355 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19480.4_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline