Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
NM_001286629.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
NM_212486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004319.mm.1 |
NM_212486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
NM_001286629.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
NM_212486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004318.mm.1 |
NM_212486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004315.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004315.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004315.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
NM_001286629.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
NM_212486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004320.mm.1 |
NM_212486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
NM_001286629.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
NM_212486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004322.mm.1 |
NM_212486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004321.mm.1 |
TC0600000564.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004321.mm.1 |
NM_001077410.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004321.mm.1 |
NM_001077410.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004321.mm.1 |
NM_212486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004321.mm.1 |
NM_212486.2 |
Refseq |