Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327594_m1
          See other CARD9 GE Assays ›
          Gene Symbol:
          Card9
          Gene Name:
          caspase recruitment domain family, member 9
          Gene Aliases:
          Gm782
          Chromosome Location:
          Chr.2: 26352236 - 26360979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327594_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Card9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037747.1 NP_001032836.1 9 - 10 1363 94
          GenBank mRNA BC151023.1 - 9 - 10 1571 9087936 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003187.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          I-kappaB kinase/NF-kappaB signaling
          response to fungus
          response to peptidoglycan
          response to muramyl dipeptide
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          response to drug
          regulation of tumor necrosis factor biosynthetic process
          regulation of apoptotic process
          response to exogenous dsRNA
          regulation of interleukin-2 biosynthetic process
          innate immune response
          positive regulation of innate immune response
          regulation of interleukin-6 biosynthetic process
          positive regulation of JNK cascade
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          protein binding
          protein domain specific binding
          protein homodimerization activity
          CARD domain binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986722 BC151023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986722 NM_001037747.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883183 AK082393.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142821 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986722 BC151023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986722 NM_001037747.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142821 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01983657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21078077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01553613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01864851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01864867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01983649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01864846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01983657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01983652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02249069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21078125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480970 - -
          Affymetrix Mouse Gene 1.1 ST Array 10480956 - -
          Affymetrix Mouse Clariom S Array PSR0200026551.mm.1 NM_001037747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026551.mm.1 NM_001037747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026551.mm.1 TC0200003187.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026549.mm.1 NM_001037747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026549.mm.1 NM_001037747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026549.mm.1 TC0200003187.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026550.mm.1 NM_001037747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026550.mm.1 NM_001037747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026550.mm.1 TC0200003187.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline