| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200009654.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009654.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009654.mm.1 |
TC0200001204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009648.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009648.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009648.mm.1 |
TC0200001204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009649.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009649.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009649.mm.1 |
TC0200001204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009650.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009650.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009650.mm.1 |
TC0200001204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009652.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009652.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009652.mm.1 |
TC0200001204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009653.mm.1 |
NM_175514.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009653.mm.1 |
NM_175514.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009653.mm.1 |
TC0200001204.mm.1 |
MTA TC |